Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl DL-2-bromopropionate, 99%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
Packaging:
Glass bottle
5445-17-0
C4H7BrO2
167
MFCD00000143
ACEONLNNWKIPTM-UHFFFAOYSA-N
methyl 2-bromopropionate,propanoic acid, 2-bromo-, methyl ester,2-bromopropionic acid methyl ester,methyl dl-2-bromopropionate,methyl2-bromopropionate,methyl alpha-bromopropionate,methyl .alpha.-bromopropionate,methyl-2-bromopropanoate,methyl 2-bromo-propionate,propionic acid, 2-bromo-, methyl ester
95576
methyl 2-bromopropanoate
CC(C(=O)OC)Br
5445-17-0
C4H7BrO2
167
MFCD00000143
ACEONLNNWKIPTM-UHFFFAOYSA-N
methyl 2-bromopropionate,propanoic acid, 2-bromo-, methyl ester,2-bromopropionic acid methyl ester,methyl dl-2-bromopropionate,methyl2-bromopropionate,methyl alpha-bromopropionate,methyl .alpha.-bromopropionate,methyl-2-bromopropanoate,methyl 2-bromo-propionate,propionic acid, 2-bromo-, methyl ester
95576
methyl 2-bromopropanoate
CC(C(=O)OC)Br
Quantity 100g
Boiling Point 143.0°C to 145.0°C (19.0mmHg)
Flash Point 51°C
Density 1.4970g/mL
Color Colorless to Yellow
Packaging Glass bottle
Refractive Index 1.4509 to 1.4530
Specific Gravity 1.497
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Linear Formula CH3CH(Br)CO2CH3
Beilstein 02, 253
Solubility Information Solubility in water: immiscible
Formula Weight 167
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material Methyl DL-2-bromopropionate, 99%
Show More Show Less

GHS H Statement
Flammable liquid and vapour.
Causes severe skin burns and eye damage.
May cause respiratory irritation.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomi

GHS Signal Word: Danger

EINECSNumber : 226-642-3

TSCA : TSCA