Alfa Aesar™ Methyl elaidate, 95%

Manufacturer: Alfa Aesar™ H3045809

Quantity 10g

 View more versions of this product

Catalog No. AAH3045809

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1937-62-8
    Molecular Formula C19H36O2
    Formula Weight 296.5
    MDL Number MFCD00066521
    Synonym Elaidic acid methyl ester; trans-9-Octadecenoic acid methyl ester
    Chemical Name or Material Methyl elaidate

    Specifications

    Formula Weight 296.5
    TSCA TSCA
    Quantity 10g
    Molecular Formula C19H36O2
    MDL Number MFCD00066521
    CAS 1937-62-8
    Synonym Elaidic acid methyl ester; trans-9-Octadecenoic acid methyl ester
    Melting Point 9° to 10°C
    Chemical Name or Material Methyl elaidate
    Assay 95%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit