Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Ethyl Ketone, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
1L
4L
500mL
Packaging:
Aluminum Bottle
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC Coated Amber Glass Bottle

Chemical Identifiers

78-93-3
C4H8O
72.107
ZWEHNKRNPOVVGH-UHFFFAOYSA-N
6569
CHEBI:28398

High quality chemicals for laboratory and specialized industrial use

Specific Gravity 1L = 0.81kg
Quantity 4L
Flash Point −9.00°C
CAS 78-93-3
Packaging Aluminum Bottle
Molecular Formula C4H8O
InChI Key ZWEHNKRNPOVVGH-UHFFFAOYSA-N
SMILES CCC(=O)C
IUPAC Name butan-2-one
Molecular Weight (g/mol) 72.107
PubChem CID 6569
ChEBI CHEBI:28398
Formula Weight 72.11
Chemical Name or Material Methyl Ethyl Ketone
Grade Baker Analyzed™ ACS Reagent, Meets ACS reagent chemical requirements
Show More Show Less