Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Ethyl Ketone, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500mL
1L
4L
Packaging:
Aluminum Bottle
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC Coated Amber Glass Bottle
78-93-3
C4H8O
72.107
ZWEHNKRNPOVVGH-UHFFFAOYSA-N
2-butanone,methyl ethyl ketone,butanone,ethyl methyl ketone,methylethyl ketone,meetco,methyl acetone,methylethylketone,3-butanone,ethylmethylketon
6569
CHEBI:28398
butan-2-one
CCC(=O)C
78-93-3
C4H8O
72.107
ZWEHNKRNPOVVGH-UHFFFAOYSA-N
2-butanone,methyl ethyl ketone,butanone,ethyl methyl ketone,methylethyl ketone,meetco,methyl acetone,methylethylketone,3-butanone,ethylmethylketon
6569
CHEBI:28398
butan-2-one
CCC(=O)C

High quality chemicals for laboratory and specialized industrial use

Quantity 500mL
Flash Point −9.00°C
Specific Gravity 1L = 0.81kg
Packaging Narrow Mouth Amber Glass Bottle
Formula Weight 72.11
Grade Meets ACS reagent chemical requirements
Chemical Name or Material Methyl Ethyl Ketone