Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl formate, 97%, pure, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
10L
Packaging:
Glass bottle
Metal drum
107-31-3
C2H4O2
60.05
MFCD00003291
TZIHFWKZFHZASV-UHFFFAOYSA-N
formic acid, methyl ester,methyl methanoate,methylformiat,formic acid methyl ester,formiate de methyle,methylformiaat,mravencan methylnaty,methylformate,caswell no. 570,hcooch3
7865
CHEBI:77699
methyl formate
COC=O
107-31-3
C2H4O2
60.05
MFCD00003291
TZIHFWKZFHZASV-UHFFFAOYSA-N
formic acid, methyl ester,methyl methanoate,methylformiat,formic acid methyl ester,formiate de methyle,methylformiaat,mravencan methylnaty,methylformate,caswell no. 570,hcooch3
7865
CHEBI:77699
methyl formate
COC=O
Viscosity 0.35 mPa.s (20°C)
Water 0.05% max.
Specific Gravity 0.968
Quantity 2.5L
Packaging Glass bottle
Flash Point −32°C
Refractive Index 1.3420 to 1.3440
Infrared Spectrum Authentic
Melting Point -100.0°C
Boiling Point 31.0°C to 33.0°C
Color Colorless
Density 0.9680g/mL
Assay Percent Range 97%
Linear Formula HCO2CH3
Beilstein 02,18
Fieser 05,446
Merck Index 15,6148
Solubility Information Solubility in water: 300g/L (20°C). Other solubilities: soluble in almost all conventional organic solvent,miscible with alcohol,soluble in ether
Formula Weight 60.05
Physical Form Liquid
Percent Purity 97%
Grade Pure
Chemical Name or Material Methyl formate, 97%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful if inhaled.
Extremely flammable liquid and vapor.

GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Take precautionary measures against static discharge.
Store in a well-ventilated place. Keep cool.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 203-481-7

RTECSNumber : LQ8925000

TSCA : TSCA