Alfa Aesar™ Methyl geranate, mixture of isomers, 94%

Manufacturer: Alfa Aesar™ A1921809

Quantity 10g

 View more versions of this product

Catalog No. AAA1921809

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1189-09-9
    Molecular Formula C11H18O2
    Formula Weight 182.26
    MDL Number MFCD00036571
    Synonym Geranic acid methyl ester; Methyl 3,7-dimethyl-2,6-octadienoate
    Chemical Name or Material Methyl geranate, mixture of isomers

    Specifications

    Formula Weight 182.26
    TSCA TSCA
    Molecular Formula C11H18O2
    MDL Number MFCD00036571
    CAS 1189-09-9
    Synonym Geranic acid methyl ester; Methyl 3,7-dimethyl-2,6-octadienoate
    Boiling Point 70°C (4mmHg)
    Quantity 10g
    Flash Point 99°C (210°F)
    Chemical Name or Material Methyl geranate, mixture of isomers
    Assay 94%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit