Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl glycolate, 95%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
96-35-5
C3H6O3
90.078
MFCD00004667
GSJFXBNYJCXDGI-UHFFFAOYSA-N
methyl glycolate,methyl hydroxyacetate,acetic acid, hydroxy-, methyl ester,glycolic acid, methyl ester,acetic acid, 2-hydroxy-, methyl ester,glycolic acid methyl ester,unii-7g7r729ej0,hydroxyacetic acid methyl ester,glycolic acid methyl,methyl2-hydroxyethanoate
66774
methyl 2-hydroxyacetate
COC(=O)CO
96-35-5
C3H6O3
90.078
MFCD00004667
GSJFXBNYJCXDGI-UHFFFAOYSA-N
methyl glycolate,methyl hydroxyacetate,acetic acid, hydroxy-, methyl ester,glycolic acid, methyl ester,acetic acid, 2-hydroxy-, methyl ester,glycolic acid methyl ester,unii-7g7r729ej0,hydroxyacetic acid methyl ester,glycolic acid methyl,methyl2-hydroxyethanoate
66774
methyl 2-hydroxyacetate
COC(=O)CO
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4160 to 1.4180 (20°C, 589nm)
Quantity 5g
Color Colorless
Density 1.184g/mL
Assay Percent Range 95%
Linear Formula HOCH2COOCH3
Formula Weight 90.08
Physical Form Liquid
Percent Purity ≥94%
Chemical Name or Material Methyl glycolate
Show More Show Less