Alfa Aesar™ Methyl hexanoate, 99%

Manufacturer: Alfa Aesar™ A1075730

Quantity 250g

 View more versions of this product

Catalog No. AAA1075730

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 106-70-7
    Molecular Formula CH3(CH2)4CO2CH3
    Formula Weight 130.19
    MDL Number MFCD00009510
    UN Number UN3272
    Beilstein 1744683
    Synonym Hexanoic acid methyl ester; Hexanoic acid methyl ester
    Chemical Name or Material Methyl hexanoate

    Specifications

    Formula Weight 130.19
    Health Hazard 3 GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    MDL Number MFCD00009510
    UN Number UN3272
    Synonym Hexanoic acid methyl ester; Hexanoic acid methyl ester
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
    Flash Point 45°C (113°F)
    Boiling Point 150° to 151°C
    Melting Point −71°C
    CAS 106-70-7
    Beilstein 1744683
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H226
    Flammable liquid and vapor.
    RTECS Number MO8401400
    Molecular Formula CH3(CH2)4CO2CH3
    Quantity 250g
    Chemical Name or Material Methyl hexanoate
    Assay 99%

    GHS H Statement
    H226
    Flammable liquid and vapor.

    GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit