Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methyl-Histone H3 (Lys9) Rabbit anti-Human, Polyclonal, Bioss
Rabbit Polyclonal Antibody
Manufacturer: Bioss BS3772R
Description
Histones are relatively small proteins that contain a high proportion of positively charged amino acids, e. g., arginine and lysine. This positive charge allows histones to bind tightly to the negatively charged DNA. There are five basic types of histone molecules called H1, H2A, H2B, H3, and H4. Histones are abundantly present in virtually all eukaryotic cells.Specifications
Methyl-Histone H3 (Lys9) | |
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
H3-4 | |
Liquid | |
Rabbit | |
IgG | |
100 μL | |
-20°C | |
Primary | |
8290 |
H3-4 | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q16695 | |
H3 histone family, member T; H3.4; H3/g; H3/t; H3FT; H3t; HIST3H3; histone 3, H3; histone cluster 3, H3; Histone H3.1t | |
KLH conjugated synthetic metylpeptide derived from human Histone H3 around metylation site of Lys9. | |
Protein A | |
RUO | |
Antibody | |
Polyclonal | |
Human |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title