Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl iso-Butyl Ketone, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500mL
4L
Packaging:
Narrow Mouth Amber Glass Bottle
108-10-1
C6H12O
100.161
NTIZESTWPVYFNL-UHFFFAOYSA-N
4-methyl-2-pentanone,methyl isobutyl ketone,isopropylacetone,isobutyl methyl ketone,hexone,mibk,2-pentanone, 4-methyl,2-methyl-4-pentanone,4-methyl-2-oxopentane,methylisobutylketon
7909
CHEBI:82344
4-methylpentan-2-one
CC(C)CC(=O)C
108-10-1
C6H12O
100.161
NTIZESTWPVYFNL-UHFFFAOYSA-N
4-methyl-2-pentanone,methyl isobutyl ketone,isopropylacetone,isobutyl methyl ketone,hexone,mibk,2-pentanone, 4-methyl,2-methyl-4-pentanone,4-methyl-2-oxopentane,methylisobutylketon
7909
CHEBI:82344
4-methylpentan-2-one
CC(C)CC(=O)C

High quality chemicals for laboratory and specialized industrial use

Formula Weight 100.16
Physical Form Liquid
Grade Meets ACS reagent chemical requirements
Density 1.318g/mL at 25°C (typical)
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 0.80kg
Flash Point 14.00°C
Boiling Point 116.5°C
Color Colorless
Quantity 4L
Chemical Name or Material Methyl iso-Butyl Ketone
Show More Show Less