Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methyl isonicotinate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
Methyl Isonicotinate induces increased walking and take-off behavior in western flower thrips, Frankliniella occidentalis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl Isonicotinate induces increased walking and take-off behavior in western flower thrips, Frankliniella occidentalis.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Methyl Isonicotinate induces increased walking and take-off behavior in western flower thrips, Frankliniella occidentalis.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Specifications
Melting Point | 8°C to 8.5°C |
Density | 1.016 |
Boiling Point | 207°C to 209°C |
Flash Point | 82°C (180°F) |
Odor | Characteristic |
Refractive Index | 1.5135 |
Quantity | 25 g |
Solubility Information | Slightly soluble in water. |
Formula Weight | 137.14 |
Percent Purity | 98% |
Show More |
Safety and Handling
Hazard Category | H227-H315-H319-H335 |
Hazard Statement | GHS H Statement H319-H227 Causes serious eye irritation. Combustible liquid. |
Precautionary Statement | P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c |
EINECSNumber | 219-456-8 |
TSCA | Yes |
Recommended Storage | Keep cold; Store under Nitrogen |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction