Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Isonicotinate 98%, ACROS Organics™

Click to view available options
Quantity:
100mL
500mL
Packaging:
Glass bottle
2459-09-8
C7H7NO2
137.14
MFCD00006427
OLXYLDUSSBULGU-UHFFFAOYSA-N
trimethyl trimellitate,trimethyl 1,2,4-benzenetricarboxylate,methyl trimellitate,trimellitic acid trimethyl ester,1,2,4-benzenetricarboxylic acid, trimethyl ester,unii-16o7dt5n66,1,2,4-benzenetricarboxylic acid trimethyl ester,1,2,4-benzenetricarboxylic acid, 1,2,4-trimethyl ester,1,2,4-trimethyl benzene-1,2,4-tricarboxylate,acmc-1ckkm
17159
trimethyl benzene-1,2,4-tricarboxylate
COC(=O)C1=CC=NC=C1
2459-09-8
C7H7NO2
137.14
MFCD00006427
OLXYLDUSSBULGU-UHFFFAOYSA-N
trimethyl trimellitate,trimethyl 1,2,4-benzenetricarboxylate,methyl trimellitate,trimellitic acid trimethyl ester,1,2,4-benzenetricarboxylic acid, trimethyl ester,unii-16o7dt5n66,1,2,4-benzenetricarboxylic acid trimethyl ester,1,2,4-benzenetricarboxylic acid, 1,2,4-trimethyl ester,1,2,4-trimethyl benzene-1,2,4-tricarboxylate,acmc-1ckkm
17159
trimethyl benzene-1,2,4-tricarboxylate
COC(=O)C1=CC=NC=C1
Specific Gravity 1.161
Flash Point 82°C
Packaging Glass bottle
Melting Point 8.5°C
Quantity 100mL
Infrared Spectrum Authentic
Refractive Index 1.5120 to 1.5140
Color Brown to Orange
Boiling Point 207.0°C to 209.0°C
Density 1.1610g/mL
Assay Percent Range 98%
Beilstein 22, 46
Merck Index 10, 5034
Formula Weight 137.14
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material Methyl isonicotinate, 98%
Show More Show Less

GHS P Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

EINECSNumber : 219-546-8

TSCA : TSCA