Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Isonicotinate 98%, ACROS Organics™

Click to view available options
Quantity:
100mL
500mL
Packaging:
Glass bottle
2459-09-8
C7H7NO2
137.14
MFCD00006427
OLXYLDUSSBULGU-UHFFFAOYSA-N
methyl isonicotinate,isonicotinic acid methyl ester,4-pyridinecarboxylic acid, methyl ester,4-picolinic acid methyl ester,4-methoxycarbonylpyridine,methylisonicotinate,methyl 4-pyridinecarboxylate,4-carbomethoxypyridine,isonicotinic acid, methyl ester,i-nicotinic acid, methyl ester
227085
methyl pyridine-4-carboxylate
COC(=O)C1=CC=NC=C1
2459-09-8
C7H7NO2
137.14
MFCD00006427
OLXYLDUSSBULGU-UHFFFAOYSA-N
methyl isonicotinate,isonicotinic acid methyl ester,4-pyridinecarboxylic acid, methyl ester,4-picolinic acid methyl ester,4-methoxycarbonylpyridine,methylisonicotinate,methyl 4-pyridinecarboxylate,4-carbomethoxypyridine,isonicotinic acid, methyl ester,i-nicotinic acid, methyl ester
227085
methyl pyridine-4-carboxylate
COC(=O)C1=CC=NC=C1
Specific Gravity 1.161
Refractive Index 1.5120 to 1.5140
Flash Point 82°C
Packaging Glass bottle
Color Brown to Orange
Density 1.1610g/mL
Boiling Point 207.0°C to 209.0°C
Quantity 500mL
Melting Point 8.5°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Beilstein 22, 46
Merck Index 10, 5034
Formula Weight 137.14
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material Methyl isonicotinate, 98%
Show More Show Less

GHS P Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

EINECSNumber : 219-546-8

TSCA : TSCA