Alfa Aesar™ Methyl isothiocyanate, 98%

Manufacturer: Alfa Aesar™ A1175736

Quantity 500g

 View more versions of this product

Catalog No. AAA1175736

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 556-61-6
    Linear Formula CH3NCS
    Formula Weight 73.12
    MDL Number MFCD00004818
    UN Number UN2477
    Beilstein 605319
    Merck Index 14,6092
    Chemical Name or Material Methyl isothiocyanate

    Specifications

    Formula Weight 73.12
    Physical Form Fused Solid
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H301-H331-H314-H317
    Toxic if swallowed.
    Toxic if inhaled.
    Causes severe skin burns and eye damage.
    May cause an allergic skin reaction.
    MDL Number MFCD00004818
    CAS 556-61-6
    Beilstein 605319
    Boiling Point 117° to 118°C
    Melting Point 31° to 36°C
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: METHYL ISOTHIOCYANATE
    UN Number UN2477
    Quantity 500g
    Flash Point 30°C (86°F)
    Storage Note 1 2°C to 8°C
    RTECS Number PA9625000
    TSCA TSCA
    Linear Formula CH3NCS
    Merck Index 14,6092
    Chemical Name or Material Methyl isothiocyanate
    Assay 98%

    GHS H Statement
    H301-H331-H314-H317
    Toxic if swallowed.
    Toxic if inhaled.
    Causes severe skin burns and eye damage.
    May cause an allergic skin reaction.

    GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    2°C to 8°C

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit