Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl isothiocyanate, 98%

Manufacturer:  Alfa Aesar™ A117570B

Quantity 1000g

 View more versions of this product

Catalog No. AAA117570B


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

556-61-6
73.12
UN2477
14,6092
CH3NCS
MFCD00004818
605319
Methyl isothiocyanate

Specifications

556-61-6
Fused Solid
73.12
UN2477
14,6092
117° to 118°C
1000g
Methyl isothiocyanate
1.069g/mL
CH3NCS
MFCD00004818
605319
White to pale yellow
31° to 36°C
Moisture Sensitive
98%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H331-H314-H317
Toxic if swallowed.
Toxic if inhaled.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: I; Proper Shipping Name: METHYL ISOTHIOCYANATE

RTECSNumber : PA9625000

TSCA : TSCA

storageNote1 : 2° to 8°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit