Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl-L-fucopyranoside, 99+%, ACROS Organics™

Click to view available options
Quantity:
500mg
Packaging:
Glass bottle
65310-00-1
C7H14O5
178.184
MFCD03427600
OHWCAVRRXKJCRB-DVEMRHSHSA-N
methyl l-fucopyranoside,methyl fucopyranoside,3s,4r,5s,6s-2-methoxy-6-methyloxane-3,4,5-triol,3s,4r,5s,6s-2-methoxy-6-methyltetrahydro-2h-pyran-3,4,5-triol,methyl 6-deoxy-galactopyranoside,methyl 6-deoxy-l-galactopyranoside,a,,a mixture,l-galactopyranoside,methyl 6-deoxy,3s,5s,6s,4r-2-methoxy-6-methyl-2h-3,4,5,6-tetrahydropyran-3,4,5-triol
2734151
(3S,4R,5S,6S)-2-methoxy-6-methyloxane-3,4,5-triol
CC1C(C(C(C(O1)OC)O)O)O
65310-00-1
C7H14O5
178.184
MFCD03427600
OHWCAVRRXKJCRB-DVEMRHSHSA-N
methyl l-fucopyranoside,methyl fucopyranoside,3s,4r,5s,6s-2-methoxy-6-methyloxane-3,4,5-triol,3s,4r,5s,6s-2-methoxy-6-methyltetrahydro-2h-pyran-3,4,5-triol,methyl 6-deoxy-galactopyranoside,methyl 6-deoxy-l-galactopyranoside,a,,a mixture,l-galactopyranoside,methyl 6-deoxy,3s,5s,6s,4r-2-methoxy-6-methyl-2h-3,4,5,6-tetrahydropyran-3,4,5-triol
2734151
(3S,4R,5S,6S)-2-methoxy-6-methyloxane-3,4,5-triol
CC1C(C(C(C(O1)OC)O)O)O
Infrared Spectrum Authentic
Color White
Packaging Glass bottle
Quantity 500mg
Specific Rotation +12.5° to +13.0° (20°C, 589nm) (C=2, Water)
Melting Point 119°C to 123°C
Assay Percent Range 99+%
Formula Weight 178.19
Physical Form Crystalline Powder
Percent Purity ≥99.0%
Chemical Name or Material Methyl-L-fucopyranoside
Show More Show Less