Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl laurate, 98+%, ACROS Organics™

Click to view available options
Quantity:
2.5g
10g
50g
250g
Packaging:
Glass bottle
111-82-0
C13H26O2
214.35
MFCD00008966
UQDUPQYQJKYHQI-UHFFFAOYSA-N
methyl laurate,lauric acid methyl ester,dodecanoic acid, methyl ester,methyl laurinate,methyl dodecylate,methyl n-dodecanoate,uniphat a40,metholene 2296,lauric acid, methyl ester,stepan c40
8139
CHEBI:87494
methyl dodecanoate
CCCCCCCCCCCC(=O)OC
111-82-0
C13H26O2
214.35
MFCD00008966
UQDUPQYQJKYHQI-UHFFFAOYSA-N
methyl laurate,lauric acid methyl ester,dodecanoic acid, methyl ester,methyl laurinate,methyl dodecylate,methyl n-dodecanoate,uniphat a40,metholene 2296,lauric acid, methyl ester,stepan c40
8139
CHEBI:87494
methyl dodecanoate
CCCCCCCCCCCC(=O)OC
Melting Point 4°C to 5°C
Boiling Point 261°C
Flash Point >110°C
Density 0.8700g/mL
Refractive Index 1.4310 to 1.4330
Specific Gravity 0.87
Packaging Glass bottle
Quantity 50g
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Linear Formula CH3(CH2)10CO2CH3
Beilstein 02,361
Solubility Information Solubility in water: insoluble
Formula Weight 214.35
Physical Form Liquid
Percent Purity 98+%
Chemical Name or Material Methyl laurate
Show More Show Less

GHS Signal Word: Warning

EINECSNumber : 203-911-3