Alfa Aesar™ Methyl malonyl chloride, 97%

Manufacturer: Alfa Aesar™ 2082314

Quantity 25g

 View more versions of this product

Catalog No. AA2082314

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 37517-81-0
    Formula Weight 136.53
    MDL Number MFCD00013657
    UN Number UN3265
    Synonym Methyl 3-chloro-3-oxopropionate; Malonic acid monomethyl ester chloride
    Chemical Name or Material Methyl malonyl chloride

    Specifications

    Formula Weight 136.53
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Synonym Methyl 3-chloro-3-oxopropionate; Malonic acid monomethyl ester chloride
    Health Hazard 2 GHS H Statement
    H314-H318-H227
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Combustible liquid.
    Physical Form Liquid
    MDL Number MFCD00013657
    CAS 37517-81-0
    Color Colorless
    Boiling Point 58°C (12mmHg)
    Quantity 25g
    Flash Point 80° (176°F)
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
    Refractive Index 1.4325
    Health Hazard 1 Danger
    Sensitivity Moisture Sensitive
    Density 1.27g/mL
    UN Number UN3265
    Chemical Name or Material Methyl malonyl chloride
    Assay 97%

    GHS H Statement
    H314-H318-H227
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Combustible liquid.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit