Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl malonyl chloride, 97%

Manufacturer:  Alfa Aesar™ 2082303

Quantity 1g

 View more versions of this product

Catalog No. AA2082303


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

37517-81-0
MFCD00013657
Methyl 3-chloro-3-oxopropionate; Malonic acid monomethyl ester chloride
136.53
UN3265
Methyl malonyl chloride

Specifications

37517-81-0
1.4325
136.53
UN3265
Methyl 3-chloro-3-oxopropionate; Malonic acid monomethyl ester chloride
1g
Moisture Sensitive
97%
1.27g/mL
Liquid
MFCD00013657
Colorless
58°C (12mmHg)
80° (176°F)
Methyl malonyl chloride

Safety and Handling

SDS & Safety Documents

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit