Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl N-benzylpyrrolidine-3-carboxylate, 97%

Manufacturer:  Alfa Aesar™ H3334106

Quantity 5g

 View more versions of this product

Catalog No. AAH3334106


  Check Availability
Add to cart

Specifications

Description

Applications
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

Solubility
Sparingly soluble in water.(0.26 g/L) (25°C),

Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.
Description & Specifications

Chemical Identifiers

17012-21-4
219.28
N-Benzylpyrrolidine-3-carboxylic acid methyl ester
C13H17NO2
MFCD04038678
Methyl N-benzylpyrrolidine-3-carboxylate

Specifications

17012-21-4
219.28
N-Benzylpyrrolidine-3-carboxylic acid methyl ester
5g
97%
C13H17NO2
MFCD04038678
98°C (0.2mmHg)
Methyl N-benzylpyrrolidine-3-carboxylate
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit