Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Nicotinate 99%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
Packaging:
Plastic bottle
93-60-7
C7H7NO2
137.14
MFCD00006388
YNBADRVTZLEFNH-UHFFFAOYSA-N
methyl nicotinate,nikomet,methylnicotinate,nicotinic acid methyl ester,nicometh,methyl 3-pyridinecarboxylate,methyl-nicotinate,3-pyridinecarboxylic acid, methyl ester,heat spray,nicotinic acid, methyl ester
7151
methyl pyridine-3-carboxylate
COC(=O)C1=CN=CC=C1
93-60-7
C7H7NO2
137.14
MFCD00006388
YNBADRVTZLEFNH-UHFFFAOYSA-N
methyl nicotinate,nikomet,methylnicotinate,nicotinic acid methyl ester,nicometh,methyl 3-pyridinecarboxylate,methyl-nicotinate,3-pyridinecarboxylic acid, methyl ester,heat spray,nicotinic acid, methyl ester
7151
methyl pyridine-3-carboxylate
COC(=O)C1=CN=CC=C1
Packaging Plastic bottle
Infrared Spectrum Authentic
Flash Point 95°C
Quantity 100g
Boiling Point 209.0°C
Melting Point 38.0°C to 43.0°C
Color Brown to White
Assay Percent Range 99%
Beilstein 22, 39
Merck Index 15, 6173
Solubility Information Solubility in water: negligible. Other solubilities: soluble in alcohol and benzene
Formula Weight 137.14
Physical Form Crystals or Crystalline Powder
Percent Purity 99%
Chemical Name or Material Methyl nicotinate, 99%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 202-261-8

RTECSNumber : QT1925000

TSCA : TSCA