Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl nonanoate, 95%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
1731-84-6
C10H20O2
172.27
MFCD00009569
IJXHLVMUNBOGRR-UHFFFAOYSA-N
methyl pelargonate,pelargonic acid methyl ester,methyl nonylate,nonanoic acid, methyl ester,methyl n-nonanoate,nonanoic acid methyl ester,unii-7xkd6qoh68,fema no. 2724,methyl ester nonanoic acid,7xkd6qoh68
15606
CHEBI:44499
methyl nonanoate
CCCCCCCCC(=O)OC
1731-84-6
C10H20O2
172.27
MFCD00009569
IJXHLVMUNBOGRR-UHFFFAOYSA-N
methyl pelargonate,pelargonic acid methyl ester,methyl nonylate,nonanoic acid, methyl ester,methyl n-nonanoate,nonanoic acid methyl ester,unii-7xkd6qoh68,fema no. 2724,methyl ester nonanoic acid,7xkd6qoh68
15606
CHEBI:44499
methyl nonanoate
CCCCCCCCC(=O)OC
Melting Point -30°C
Specific Gravity 0.874
Boiling Point 213°C
Flash Point 81°C
Refractive Index 1.4210 to 1.423
Color Colorless
Infrared Spectrum Authentic
Quantity 5g
Packaging Glass bottle
Density 0.8740g/mL
Assay Percent Range 94% min. (GC)
Linear Formula CH3(CH2)7C(O)OCH3
Solubility Information Solubility in water: practically insoluble
Formula Weight 172.27
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material Methyl nonanoate
Show More Show Less

GHS H Statement:
Causes skin irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 217-052-7