Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl o-toluate, 99%

Click to view available options
Quantity:
25g
100g
500g
89-71-4
C9H10O2
150.177
MFCD00008428
WVWZECQNFWFVFW-UHFFFAOYSA-N
methyl o-toluate,methyl orthotoluate,o-toluic acid, methyl ester,methyl o-methylbenzoate,methyl toluate,benzoic acid, 2-methyl-, methyl ester,2-methylbenzoic acid methyl ester,o-toluic acid methyl ester,methyl-o-methyl benzoate,2-methyl methylbenzoate
33094
methyl 2-methylbenzoate
CC1=CC=CC=C1C(=O)OC
89-71-4
C9H10O2
150.177
MFCD00008428
WVWZECQNFWFVFW-UHFFFAOYSA-N
methyl o-toluate,methyl orthotoluate,o-toluic acid, methyl ester,methyl o-methylbenzoate,methyl toluate,benzoic acid, 2-methyl-, methyl ester,2-methylbenzoic acid methyl ester,o-toluic acid methyl ester,methyl-o-methyl benzoate,2-methyl methylbenzoate
33094
methyl 2-methylbenzoate
CC1=CC=CC=C1C(=O)OC
Applications
It has been used in the preparation of 2-(azidomethyl)benzoic acid.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep ignition sources away.
Flash Point 82°C (179°F)
Density 1.072
Refractive Index 1.519
Boiling Point 207°C to 208°C
Quantity 25g
Beilstein 2041846
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 150.18
Percent Purity 99%
Chemical Name or Material Methyl o-toluate

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

EINECSNumber : 201-932-2

TSCA : Yes

Recommended Storage : Ambient temperatures