Alfa Aesar™ Methyl octanoate, 99%

Manufacturer: Alfa Aesar™ A109910B

Quantity 1000g

 View more versions of this product

Catalog No. AAA109910B

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 111-11-5
    Molecular Formula CH3(CH2)6CO2CH3
    Formula Weight 158.24
    MDL Number MFCD00009551
    Beilstein 1752270
    Synonym Methyl caprylate; Octanoic acid methyl ester
    Chemical Name or Material Methyl octanoate

    Specifications

    Formula Weight 158.24
    Quantity 1000g
    TSCA TSCA
    CAS 111-11-5
    Flash Point 72°C (161°F)
    Boiling Point 193° to 194°C
    Molecular Formula CH3(CH2)6CO2CH3
    MDL Number MFCD00009551
    Beilstein 1752270
    Synonym Methyl caprylate; Octanoic acid methyl ester
    Melting Point −40°C
    RTECS Number RH0778000
    Chemical Name or Material Methyl octanoate
    Assay 99%

    Description

    Applications
    Methyl octanoate-ethanol mixtures constitute the biodiesel-bioethanol surrogate fuel and kinetics of its oxidation has been studied experimentally in a jet-stirred reactor. Deoxygenation of methyl octanoate over alumina-supported Pt has been studied in both the vapor phase in a flow reactor and in the liquid phase in a semi-batch reactor. It is also used to make other chemicals.

    Solubility
    Insoluble in water.

    Notes
    Stable under recommended storage conditions. Incompatible with oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit