Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Orange, ACS Reagent (Acid Orange 52. CI 13025), Reagents

Catalog No. p-200022043
Click to view available options
Quantity:
25 g
100 g
500 g
25 kg

Methyl Orange, ACS Reagent (Acid Orange 52. CI 13025), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 547-58-0
Molecular Formula C14H14N3O3S
Molecular Weight (g/mol) 304.34
InChI Key IETWCRRCPURZOC-UHFFFAOYSA-M
Synonym Orange III, Helianthine
IUPAC Name 4-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzene-1-sulfonate
SMILES CN(C)C1=CC=C(C=C1)N=NC1=CC=C(C=C1)S([O-])(=O)=O

Specifications

Grade ACS
Packaging Glass Bottle
Specific Gravity 0.987
Color Orange to Yellow
Quantity 100 g
Physical Form Solid
Chemical Name or Material Methyl Orange, ACS Reagent (Acid Orange 52. CI 13025)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.