Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Orange Indicator, 0.1% w/v Prepared in accordance with ASTM D974 procedures, Reagents

Catalog No. 01336740
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-336-740 500 mL
01-336-744 120 mL
01-336-743 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-336-740 Supplier Reagents Holdings Llc Supplier No. C6118000500A
Only null left
Add to Cart
Add to Cart

Methyl Orange Indicator, 0.1% w/v Prepared in accordance with ASTM D974 procedures, ASTM, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Methyl Orange Indicator, 0.1% w/v Prepared in accordance with ASTM D974 procedures
CAS 7732-18-5, 547-58-0
Boiling Point 100°C
Molecular Formula C14H14N3NaO3S/H2O
Synonym Orange III, Helianthine
Percent Purity 0.1%
Grade ASTM
Packaging Poly Bottle
Specific Gravity 1.001
Color Orange
Quantity 500 mL
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.