Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methyl Orange Indicator, 0.1% w/v Prepared in accordance with ASTM D974 procedures, Reagents
Click to view available options
Quantity:
120 mL
500 mL
1 L
4 L
10 L
20 L
Specifications
Specifications
Chemical Name or Material | Methyl Orange Indicator, 0.1% w/v Prepared in accordance with ASTM D974 procedures |
CAS | 7732-18-5, 547-58-0 |
Boiling Point | 100°C |
Molecular Formula | C14H14N3NaO3S/H2O |
Synonym | Orange III, Helianthine |
Percent Purity | 0.1% |
Grade | ASTM |
Packaging | Cubitainer |
Specific Gravity | 1.001 |
Color | Orange |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction