Learn More
Methyl Orange Indicator, 0.1% w/v Prepared in accordance with ASTM D974 procedures, Reagents
Specifications
Specifications
| Chemical Name or Material | Methyl Orange Indicator, 0.1% w/v Prepared in accordance with ASTM D974 procedures |
| CAS | 7732-18-5, 547-58-0 |
| Boiling Point | 100°C |
| Molecular Formula | C14H14N3NaO3S/H2O |
| Synonym | Orange III, Helianthine |
| Percent Purity | 0.1% |
| Grade | ASTM |
| Packaging | Poly Bottle |
| Specific Gravity | 1.001 |
| Color | Orange |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.