Methyl Orange Indicator Solution, Modified (Methyl Orange-Xylene Cyanol Mixed Indicator), Ricca Chemical

Manufacturer: Ricca Chemical CompanySupplier Diversity Partner 50304

Packaging Glass amber
Quantity 120mL

 View more versions of this product

Catalog No. 5030-4

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 547-58-0 (Methyl Orange, Sodium Salt),2650-17-1 (Xylene Cyanol FF),7732-18-5 (Water, Deionized)
    Physical Form Liquid
    Chemical Name or Material Methyl Orange Indicator Solution, (Methyl Orange-Xylene Cyanol Mixed Indicator)

    Specifications

    Physical Form Liquid
    Color Clear green
    Quantity 120mL
    Treatment(s) Modified
    Packaging Glass amber
    CAS 2650-17-1 (Xylene Cyanol FF),547-58-0 (Methyl Orange, Sodium Salt),7732-18-5 (Water, Deionized)
    Chemical Name or Material Methyl Orange Indicator Solution, (Methyl Orange-Xylene Cyanol Mixed Indicator)

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit