Methyl Orange, Sodium Salt, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

Manufacturer: J.T.Baker 269400

Packaging Wide Mouth Amber Glass Bottle
Quantity 30g

 View more versions of this product

Catalog No. 02-003-486

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 

  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 547-58-0
    Molecular Formula 4-(CH3)2NC6H4N:NC6H4-4-SO3Na
    Formula Weight 327.34
    Synonym C.I. 13025
    Chemical Name or Material Methyl Orange, Sodium Salt
    Grade Baker Analyzed™ ACS Reagent

    Specifications

    Formula Weight 327.34
    Physical Form Powder
    Molecular Formula 4-(CH3)2NC6H4N:NC6H4-4-SO3Na
    Packaging Wide Mouth Amber Glass Bottle
    Specific Gravity 1L = 1.00kg
    Quantity 30g
    CAS 547-58-0
    Synonym C.I. 13025
    Chemical Name or Material Methyl Orange, Sodium Salt
    Grade Baker Analyzed™ ACS Reagent

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit