Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methyl Orange solution, Honeywell Fluka™
Click to view available options
Quantity:
500 mL
1 L
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
UN Number | UN1993 |
Beilstein | 4732884 |
Formula Weight | 327.33g/mol |
Percent Purity | 0.10% |
Quantity | 1 L |
Flash Point | 39°C (102.2°F) |
Chemical Name or Material | Methyl Orange solution |
Safety and Handling
Hazard Statement | P210-P403 + P235 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction