Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Orange solution, Honeywell Fluka™

Click to view available options
Quantity:
500mL
1L
6 x 500mL
6 x 1L
547-58-0
C14H14N3NaO3S
327.334
MFCD00007502
STZCRXQWRGQSJD-UHFFFAOYSA-M
23673835
sodium;4-[[4-(dimethylamino)phenyl]diazenyl]benzenesulfonate
CN(C)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)S(=O)(=O)[O-].[Na+]
547-58-0
C14H14N3NaO3S
327.334
MFCD00007502
STZCRXQWRGQSJD-UHFFFAOYSA-M
23673835
sodium;4-[[4-(dimethylamino)phenyl]diazenyl]benzenesulfonate
CN(C)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)S(=O)(=O)[O-].[Na+]

Indicator, 0.1%, Reag. Ph. Eur.

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
UN Number UN1993
Beilstein 4732884
Formula Weight 327.33g/mol
Percent Purity 0.10%
Quantity 500mL
Flash Point 39°C (102.2°F)
Chemical Name or Material Methyl Orange solution

P210-P403 + P235