Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methyl Orange-Xylene Cyanole Indicator, in 50% v/v Methanol, Reagents
Click to view available options
Quantity:
500 mL
Specifications
Specifications
| Chemical Name or Material | Methyl Orange-Xylene Cyanole Indicator, in 50% v/v Methanol |
| Boiling Point | 65°C |
| CAS | 7732-18-5, 67-56-1, 2650-17-1 |
| Synonym | Modified Methyl Orange |
| Grade | Reagent |
| Packaging | Poly Bottle |
| Specific Gravity | 0.8965 |
| Color | Blue to Green |
| Quantity | 500 mL |
| Physical Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction