Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methyl Orange-Xylene Cyanole Indicator, Reagents
Click to view available options
Quantity:
120 mL
500 mL
1 L
4 L
Specifications
Specifications
Chemical Name or Material | Methyl Orange-Xylene Cyanole Indicator |
CAS | 7732-18-5, 2650-17-1, 2650-17-1 |
Boiling Point | 100°C |
Synonym | Modified Methyl Orange |
Grade | Reagent |
Packaging | Poly Bottle |
Specific Gravity | 1.0005 |
Color | Blue to Green |
Quantity | 120 mL |
Physical Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction