Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Methyl-p-benzoquinone, 98%
Click to view available options
Quantity:
10g
50g
Specifications
Quantity | 50g |
Melting Point | 67°C to 70°C |
Color | Green-Yellow |
Solubility Information | Insoluble in water. |
Formula Weight | 122.12 |
Physical Form | Crystalline |
Percent Purity | 98% |
Chemical Name or Material | Methyl-p-benzoquinone |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H301-H315-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 209-056-2
RTECSNumber : DK6300000
TSCA : Yes
Recommended Storage : Ambient temperatures