Alfa Aesar™ Methyl p-tolyl sulfone, 99%

Manufacturer: Alfa Aesar™ A1153114

Quantity 25g

 View more versions of this product

Catalog No. AAA1153114

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3185-99-7
    Molecular Formula C8H10O2S
    Formula Weight 170.23
    MDL Number MFCD00014742
    Beilstein 607296
    Synonym 4-(Methylsulfonyl)toluene
    Chemical Name or Material Methyl p-tolyl sulfone

    Specifications

    Formula Weight 170.23
    Molecular Formula C8H10O2S
    MDL Number MFCD00014742
    Beilstein 607296
    Synonym 4-(Methylsulfonyl)toluene
    Quantity 25g
    CAS 3185-99-7
    Melting Point 86° to 89°C
    Boiling Point 140°C (3mmHg)
    Chemical Name or Material Methyl p-tolyl sulfone
    Assay 99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit