Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl pentafluoropropionate, 98%

Click to view available options
Quantity:
5g
25g
378-75-6
C4H3F5O2
178.058
MFCD00000430
JMKJCPUVEMZGEC-UHFFFAOYSA-N
methyl pentafluoropropionate,methyl perfluoropropionate,methylpentafluoropropionate,pentafluoropropionic acid methyl ester,propanoic acid, pentafluoro-, methyl ester,methyl pentafluoropropanoate,perfluoropropionic acid, methyl ester,pentafluoropropanoic acid methyl ester,2,2,3,3,3-pentafluoro-propionic acid methyl ester,methylester kyseliny pentafluorpropionove
9783
methyl 2,2,3,3,3-pentafluoropropanoate
COC(=O)C(C(F)(F)F)(F)F
378-75-6
C4H3F5O2
178.058
MFCD00000430
JMKJCPUVEMZGEC-UHFFFAOYSA-N
methyl pentafluoropropionate,methyl perfluoropropionate,methylpentafluoropropionate,pentafluoropropionic acid methyl ester,propanoic acid, pentafluoro-, methyl ester,methyl pentafluoropropanoate,perfluoropropionic acid, methyl ester,pentafluoropropanoic acid methyl ester,2,2,3,3,3-pentafluoro-propionic acid methyl ester,methylester kyseliny pentafluorpropionove
9783
methyl 2,2,3,3,3-pentafluoropropanoate
COC(=O)C(C(F)(F)F)(F)F
Density 1.393
Refractive Index 1.288
Boiling Point 59°C to 60°C
Flash Point −1°C (30°F)
Quantity 5g
UN Number UN3272
Beilstein 1777501
Formula Weight 178.06
Percent Purity 98%
Chemical Name or Material Methyl pentafluoropropionate

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H225-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ESTERS, N.O.S.

EINECSNumber : 206-828-0

RTECSNumber : UF6475500

TSCA : Yes

Recommended Storage : Ambient temperatures