Alfa Aesar™ Methyl perfluorodecanoate, 90+%

Manufacturer: Alfa Aesar™ B22238-06

Quantity 5g

 View more versions of this product

Catalog No. AAB2223806

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 307-79-9
    Molecular Formula CF3(CF2)8CO2CH3
    Formula Weight 528.11g/mol
    MDL Number MFCD00153211
    Beilstein 1810875
    Synonym Methyl nonadecafluorodecanoate; Perfluorodecanoic acid methyl ester
    Chemical Name or Material Methyl perfluorodecanoate

    Specifications

    Boiling Point 193°C
    Quantity 5g
    Flash Point 81°C (177°F)
    CAS 307-79-9
    Molecular Formula CF3(CF2)8CO2CH3
    Formula Weight 528.11g/mol
    MDL Number MFCD00153211
    Beilstein 1810875
    Synonym Methyl nonadecafluorodecanoate; Perfluorodecanoic acid methyl ester
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H227
    Combustible liquid.
    Health Hazard 3 GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material Methyl perfluorodecanoate
    Assay ≥90%

    GHS H Statement
    H227
    Combustible liquid.

    GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit