Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Methyl perfluorooctanoate, 97%
Click to view available options
Quantity:
5g
25g
Specifications
Refractive Index | 1.305 |
Density | 1.684 |
Quantity | 5g |
Flash Point | 65°C (149°F) |
Boiling Point | 158°C to 159°C |
Beilstein | 1809817 |
Formula Weight | 428.09 |
Percent Purity | 97% |
Chemical Name or Material | Methyl perfluorooctanoate |
Safety and Handling
GHS H Statement
H227
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
EINECSNumber : 206-808-1
RTECSNumber : RH0783200
TSCA : Yes
Recommended Storage : Ambient temperatures