Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl perfluorooctanoate, 97%

Click to view available options
Quantity:
5g
25g
376-27-2
C9H3F15O2
428.097
MFCD00039244
XOCNYZFAMHDXJK-UHFFFAOYSA-N
methyl perfluorooctanoate,methyl pentadecafluorooctanoate,pentadecafluorooctanoic acid, methyl ester,methyl perfluorocaprylate,octanoic acid, pentadecafluoro-, methyl ester,perfluorooctanoic acid methyl ester,pentadecafluorooctanoic acid methyl ester,octanoic acid, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluoro-, methyl ester,acmc-1cndk,methylpentadecafluorooctanoate
67824
methyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctanoate
COC(=O)C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
376-27-2
C9H3F15O2
428.097
MFCD00039244
XOCNYZFAMHDXJK-UHFFFAOYSA-N
methyl perfluorooctanoate,methyl pentadecafluorooctanoate,pentadecafluorooctanoic acid, methyl ester,methyl perfluorocaprylate,octanoic acid, pentadecafluoro-, methyl ester,perfluorooctanoic acid methyl ester,pentadecafluorooctanoic acid methyl ester,octanoic acid, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluoro-, methyl ester,acmc-1cndk,methylpentadecafluorooctanoate
67824
methyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctanoate
COC(=O)C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
Flash Point 65°C (149°F)
Quantity 25g
Boiling Point 158°C to 159°C
Beilstein 1809817
Formula Weight 428.09
Percent Purity 97%
Chemical Name or Material Methyl perfluorooctanoate

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : RH0783200

TSCA : TSCA