Alfa Aesar™ Methyl phenyl sulfone, 98+%

Manufacturer: Alfa Aesar™ A1172718

Quantity 50g

 View more versions of this product

Catalog No. AAA1172718

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3112-85-4
    Molecular Formula C7H8O2S
    Formula Weight 156.2
    MDL Number MFCD00014741
    Beilstein 1906914
    Chemical Name or Material Methyl phenyl sulfone

    Specifications

    Formula Weight 156.2
    Health Hazard 3 GHS P Statement
    P264-P270-P301+P312-P330-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00014741
    CAS 3112-85-4
    Health Hazard 1 Warning
    RTECS Number WR6800000
    Molecular Formula C7H8O2S
    Beilstein 1906914
    Health Hazard 2 GHS H Statement
    H302
    Harmful if swallowed.
    Melting Point 85° to 88°C
    Quantity 50g
    Chemical Name or Material Methyl phenyl sulfone
    Assay ≥98%

    Description

    Applications
    Methyl phenyl sulfone, a versatile building block for the construction of chemicals with diverse functionalities

    Solubility
    Soluble in methanol, ethanol, benzene, and chloroform. Partly miscible in water.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

    GHS H Statement
    H302
    Harmful if swallowed.

    GHS P Statement
    P264-P270-P301+P312-P330-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit