Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl phenyl sulfone, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
3112-85-4
C7H8O2S
156.2
MFCD00014741
JCDWETOKTFWTHA-UHFFFAOYSA-N
methyl phenyl sulfone,methylsulfonyl benzene,phenyl methyl sulfone,benzene, methylsulfonyl,sulfone, methyl phenyl,phenylsulfonyl methane,methylphenylsulfone,methylsulphonyl benzene,methanesulfonylbenzene,methyl phenyl sulphone
18369
methylsulfonylbenzene
CS(=O)(=O)C1=CC=CC=C1
3112-85-4
C7H8O2S
156.2
MFCD00014741
JCDWETOKTFWTHA-UHFFFAOYSA-N
methyl phenyl sulfone,methylsulfonyl benzene,phenyl methyl sulfone,benzene, methylsulfonyl,sulfone, methyl phenyl,phenylsulfonyl methane,methylphenylsulfone,methylsulphonyl benzene,methanesulfonylbenzene,methyl phenyl sulphone
18369
methylsulfonylbenzene
CS(=O)(=O)C1=CC=CC=C1
Infrared Spectrum Authentic
Melting Point 85°C to 88°C
Quantity 25g
Color White
Packaging Glass bottle
Assay Percent Range 98.5% min. (GC)
Linear Formula CH3SO2C6H5
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol,benzene and chloroform,slightly soluble in carbon tetrachloride
Formula Weight 156.2
Physical Form Crystals
Percent Purity 99%
Chemical Name or Material Methyl phenyl sulfone, 99%
Show More Show Less

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 221-476-8

RTECSNumber : WR6800000