Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl propyl disulfide, 80%, Tech., ACROS Organics™

Click to view available options
Quantity:
1g
5g
2179-60-4
C4H10S2
122.244
PUCHCUYBORIUSM-UHFFFAOYSA-N
methyl propyl disulfide,disulfide, methyl propyl,1-methyldisulfanyl propane,2,3-dithiahexane,methyl propyl disulphide,methyldithiopropane,propyl methyl disulfide,methyl n-propyl disulfide,unii-8m12k3fu0t,methylpropyldisulfide
16592
1-(methyldisulfanyl)propane
CCCSSC
2179-60-4
C4H10S2
122.244
PUCHCUYBORIUSM-UHFFFAOYSA-N
methyl propyl disulfide,disulfide, methyl propyl,1-methyldisulfanyl propane,2,3-dithiahexane,methyl propyl disulphide,methyldithiopropane,propyl methyl disulfide,methyl n-propyl disulfide,unii-8m12k3fu0t,methylpropyldisulfide
16592
1-(methyldisulfanyl)propane
CCCSSC
Density 0.98g/mL
Infrared Spectrum Authentic
Packaging Glass bottle
Color Yellow
Quantity 1g
Refractive Index 1.5050 to 1.5090 (20°C, 589nm)
Assay Percent Range 80%
Linear Formula CH3CHCHSSCH3
Formula Weight 122.25
Physical Form Liquid
Percent Purity ≥75%
Grade Technical
Chemical Name or Material Methyl propyl disulfide, tech., 80%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 218-551-2

TSCA : TSCA