Alfa Aesar™ Methyl pyrazine-2-carboxylate, 98+%

Manufacturer: Alfa Aesar™ A1202806

Quantity 5g

 View more versions of this product

Catalog No. AAA1202806

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6164-79-0
    Molecular Formula C6H6N2O2
    Formula Weight 138.13
    MDL Number MFCD00014611
    Beilstein 118345
    Merck Index 14,7959
    Synonym Methyl pyrazinoate; Pyrazine-2-carboxylic acid methyl ester
    Chemical Name or Material Methyl pyrazine-2-carboxylate

    Specifications

    Formula Weight 138.13
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00014611
    Quantity 5g
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Molecular Formula C6H6N2O2
    Merck Index 14,7959
    Synonym Methyl pyrazinoate; Pyrazine-2-carboxylic acid methyl ester
    Melting Point 56° to 60°C
    TSCA TSCA
    CAS 6164-79-0
    Health Hazard 1 Warning
    Beilstein 118345
    Flash Point >100°C (212°F)
    Chemical Name or Material Methyl pyrazine-2-carboxylate
    Assay ≥98%

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit