Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl pyridazine-4-carboxylate, 96%

Manufacturer:  Alfa Aesar™ H2744906

Quantity 5g

 View more versions of this product

Catalog No. AAH2744906


  Check Availability
Add to cart

Specifications

Description

Applications
It is used to produce 4-Pyridazincarbohydroxamsaeure by heating and requires a reagent hydroxylammoniumchloride and Na2CO3. Meanwhile, it needs solvent methanol.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Description & Specifications

Chemical Identifiers

34231-77-1
138.13
Pyridazine-4-carboxylic acid methyl ester
C6H6N2O2
MFCD09953488
Methyl pyridazine-4-carboxylate

Specifications

34231-77-1
138.13
Pyridazine-4-carboxylic acid methyl ester
5g
96%
C6H6N2O2
MFCD09953488
64° to 67°C
Methyl pyridazine-4-carboxylate
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit