Methyl pyruvate, 98%

Manufacturer: Alfa Aesar A1396614

Quantity 25g

 View more versions of this product

Catalog No. AAA1396614

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 600-22-6
    Molecular Formula CH3C(O)C2CH3
    Formula Weight 102.09
    MDL Number MFCD00008754
    UN Number UN3272
    Beilstein 1361953
    Merck Index 14,8021
    Chemical Name or Material Methyl Pyruvate

    Specifications

    Formula Weight 102.09
    Quantity 25g
    MDL Number MFCD00008754
    UN Number UN3272
    Beilstein 1361953
    Merck Index 14,8021
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H226
    Flammable liquid and vapour.
    Health Hazard 3 GHS P Statement
    P261
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Recommended Storage 2° to 8°C
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
    Physical Form Liquid
    Density 1.123
    Assay Percent Range 98%
    Molecular Formula CH3C(O)C2CH3
    CAS 600-22-6
    Chemical Name or Material Methyl Pyruvate
    Assay 98%

    Description

    Applications
    Methyl Pyruvate acts as a potent secretagogue used in the study of stimulus-secretion coupling. Also used in the preparation of quinoxalines as dual phoosphodiesterase 2/10 (PDE2/10) inhibitors. It increases free calcium ion concentration in the cytosol of pancreatic cells. It is used in the preparation of dimethyl 2,3-dimethylenebutanedioate.

    Solubility
    Soluble in chloroform, methanol, ether, alcohol. Slightly soluble in water.

    Notes
    Store away from oxidizing agent, heat.

    GHS H Statement
    H226
    Flammable liquid and vapour.

    2° to 8°C

    GHS P Statement
    P261
    Avoid breathing dust/fume/gas/mist/vapors/spray.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit