Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl quinoxaline-6-carboxylate, 98%

Click to view available options
Quantity:
1g
5g
23088-23-5
C10H8N2O2
188.186
MFCD00102618
VFPWJISMXACHIG-UHFFFAOYSA-N
methyl 6-quinoxalinecarboxylate,6-quinoxalinecarboxylic acid, methyl ester,maybridge1_006081,acmc-1cnh9,methyl6-quinoxalinecarboxylate,methyl quinoxaline-7-carboxylate,6-quinoxalinecarboxylic acid methyl ester,6-quinoxalinecarboxylicacid, methyl ester,6-quinoxalinecarboxylic acid methyl ester;
2781239
methyl quinoxaline-6-carboxylate
COC(=O)C1=CC2=NC=CN=C2C=C1
23088-23-5
C10H8N2O2
188.186
MFCD00102618
VFPWJISMXACHIG-UHFFFAOYSA-N
methyl 6-quinoxalinecarboxylate,6-quinoxalinecarboxylic acid, methyl ester,maybridge1_006081,acmc-1cnh9,methyl6-quinoxalinecarboxylate,methyl quinoxaline-7-carboxylate,6-quinoxalinecarboxylic acid methyl ester,6-quinoxalinecarboxylicacid, methyl ester,6-quinoxalinecarboxylic acid methyl ester;
2781239
methyl quinoxaline-6-carboxylate
COC(=O)C1=CC2=NC=CN=C2C=C1
Quantity 1g
Melting Point 96°C to 99°C
Formula Weight 188.19
Percent Purity 98%
Chemical Name or Material Methyl quinoxaline-6-carboxylate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : No

Recommended Storage : Ambient temperatures