Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl (R)-(+)-3-(Acetylthio)-2-methylpropionate 98.0+%, TCI America™

Click to view available options
Quantity:
5mL
86961-07-1
C7H12O3S
176.23
MFCD00671491
ODXYWRPJDYJIPT-YFKPBYRVSA-N
(R)-(+)-3-(Acetylthio)-2-methylpropionic Acid Methyl Ester
10910099
methyl (2R)-3-acetylsulfanyl-2-methylpropanoate
CC(CSC(=O)C)C(=O)OC
86961-07-1
C7H12O3S
176.23
MFCD00671491
ODXYWRPJDYJIPT-YFKPBYRVSA-N
(R)-(+)-3-(Acetylthio)-2-methylpropionic Acid Methyl Ester
10910099
methyl (2R)-3-acetylsulfanyl-2-methylpropanoate
CC(CSC(=O)C)C(=O)OC
Color Yellow
Quantity 5mL
Formula Weight 176.23
Physical Form Clear Liquid at 20°C
Percent Purity ≥98.0% (GC)
Chemical Name or Material Methyl (R)-(+)-3-(Acetylthio)-2-methylpropionate