Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Red, 0.02% in Methanol, pH 4.2 to 6.3 Pink to Yellow, Certified, LabChem™

Catalog No. p-5389059
Change view
Click to view available options
Quantity:
125 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC171007 125 mL
LC171002 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC171007 Supplier LabChem, Inc. Supplier No. LC171007
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Methyl Red
Melting Point -98°C
Name Note 0.02% in Methanol, pH 4.2 to 6.3 Pink to Yellow
Boiling Point 65°C
CAS Max % 99.98, 0.02
CAS 845-10-3
Molecular Formula CH4O
Linear Formula C15H14N3O2Na
MDL Number MFCD00004595
Solubility Information Soluble in water
Identification Passes Test
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
SMILES CO
IUPAC Name methanol
Molecular Weight (g/mol) 32.04
Formula Weight 291.28
Grade Certified
Packaging Poly Bottle
Specific Gravity 0.79g/mL
Decomposition Information Carbon monoxide; Carbon dioxide; May release flammable gases
Density 0.79g/mL
Color Orange
Quantity 125 mL
Flash Point 9.7°C
Physical Form Liquid
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.