Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methyl Red Indicator, 0.02% (w/v), Ricca Chemical
$47.00 - $187.00
Specifications
Chemical Name or Material | Methyl Red Indicator |
---|---|
CAS | 845-10-3, 7732-18-5 |
Concentration | 0.02% (w/v) |
Packaging | Natural Poly Bottle |
Solvent or Matrix | Aqueous Solution |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
5050-4
|
Ricca Chemical Company
50504 |
120 mL | Natural Poly Bottle |
Each for $47.00
|
|
||||
5050-16
|
Ricca Chemical Company
505016 |
500 mL | Natural Poly Bottle |
Each for $62.50
|
|
||||
5050-32
|
Ricca Chemical Company
505032 |
1 L | Natural Poly Bottle |
Each for $187.00
|
|
||||
Specifications
Methyl Red Indicator | |
0.02% (w/v) | |
Aqueous Solution | |
Liquid |
845-10-3, 7732-18-5 | |
Natural Poly Bottle | |
Orange to Red |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title